Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCAAGGAAGAGAGAATCGCTT[A/G]AGCCCAAGACTGCAGGGCATTATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605974 MIM: 615253 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLU7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLU7 - SLU7 homolog, splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006425.4 | 3115 | UTR 3 | NP_006416.3 | |||
XM_005265793.3 | 3115 | UTR 3 | XP_005265850.1 | |||
XM_011534418.2 | 3115 | Intron | XP_011532720.1 |
ZBED8 - zinc finger BED-type containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |