Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGGAGAGCTTCAGTCCGGCTGA[C/T]GTGGCACCAGGGCCAATCTGCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190198 MIM: 612854 | ||||||||||||||||||||
Literature Links: |
C9orf163 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf163 - chromosome 9 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152571.2 | 1227 | Silent Mutation | GAC,GAT | D,D 91 | NP_689784.1 |
NOTCH1 - notch 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |