Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTCTACTTGGATCACTAGAAG[A/G]TTGCAGCTTCCATTAAAGAGGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614764 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KATNAL1 - katanin catalytic subunit A1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014380.2 | 5391 | UTR 3 | NP_001014402.1 | |||
NM_032116.4 | 5391 | UTR 3 | NP_115492.1 | |||
XM_005266574.2 | 5391 | UTR 3 | XP_005266631.1 | |||
XM_017020791.1 | 5391 | UTR 3 | XP_016876280.1 | |||
XM_017020792.1 | 5391 | UTR 3 | XP_016876281.1 | |||
XM_017020793.1 | 5391 | UTR 3 | XP_016876282.1 |