Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATGTCATTGAGCAGAATGAGAA[C/T]CACGTACTGATAAATGCAGCAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611626 MIM: 612408 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPHOSPH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPHOSPH8 - M-phase phosphoprotein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSPC1 - paraspeckle component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042414.2 | 2949 | Intron | NP_001035879.1 | |||
XM_006719844.3 | 2949 | Intron | XP_006719907.1 | |||
XM_011535137.1 | 2949 | Intron | XP_011533439.1 | |||
XM_011535138.2 | 2949 | UTR 3 | XP_011533440.1 | |||
XM_011535139.1 | 2949 | Intron | XP_011533441.1 | |||
XM_011535140.1 | 2949 | Intron | XP_011533442.1 | |||
XM_011535141.2 | 2949 | Intron | XP_011533443.1 | |||
XM_011535142.2 | 2949 | Intron | XP_011533444.1 | |||
XM_017020650.1 | 2949 | Intron | XP_016876139.1 |