Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGAAGAGTATCTCAGCTGGGAA[A/G]GACCGGGGCTCCCAGATTTCGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNA1 - cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111045.1 | 429 | Missense Mutation | AGA,GGA | R,G 27 | NP_001104515.1 | |
NM_001111046.1 | 429 | Intron | NP_001104516.1 | |||
NM_001111047.1 | 429 | Intron | NP_001104517.1 | |||
NM_003914.3 | 429 | Missense Mutation | AGA,GGA | R,G 27 | NP_003905.1 | |
XM_011535294.2 | 429 | Intron | XP_011533596.1 | |||
XM_011535295.2 | 429 | Intron | XP_011533597.1 | |||
XM_011535296.2 | 429 | Intron | XP_011533598.1 |