Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAGCTGTTTTCCTTTTATGAA[A/C]CTTTCATTTTGCTTATCTTTTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604565 MIM: 606019 MIM: 613417 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG5 - ALG5, dolichyl-phosphate beta-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EXOSC8 - exosome component 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181503.2 | Intron | NP_852480.1 | ||||
XM_006719763.1 | Intron | XP_006719826.1 |
SUPT20H - SPT20 homolog, SAGA complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |