Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGTGCCTCATAAATGCAGGGCC[C/T]CAGAGTACTCAGAAAGGGATTAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYL1 - crystallin lambda 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015974.2 | 1250 | UTR 3 | NP_057058.2 | |||
XM_005266415.3 | 1250 | UTR 3 | XP_005266472.1 | |||
XM_005266416.4 | 1250 | Intron | XP_005266473.1 | |||
XM_017020624.1 | 1250 | UTR 3 | XP_016876113.1 |