Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGCGGGCGCCCCGAGGGCGGGC[A/G]GGCGCCCCGCGCGGCCGCGGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607587 | ||||||||||||||||||||
Literature Links: |
IL17D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL17D - interleukin 17D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138284.1 | 175 | Missense Mutation | AGG,GGG | R,G 23 | NP_612141.1 | |
XM_005266421.3 | 175 | Missense Mutation | AGG,GGG | R,G 30 | XP_005266478.1 | |
XM_005266422.2 | 175 | Intron | XP_005266479.1 |