Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTAAAGATGTGAGCTTCAAGCGG[A/G]TAATGGGCAAGCCACACCTCCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1AKMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEF1AKMT1 - eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPO4 - exportin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022459.4 | 7792 | UTR 3 | NP_071904.4 | |||
XM_005266502.1 | 7792 | UTR 3 | XP_005266559.1 | |||
XM_005266503.3 | 7792 | UTR 3 | XP_005266560.1 | |||
XM_005266504.3 | 7792 | UTR 3 | XP_005266561.1 | |||
XM_011535197.1 | 7792 | UTR 3 | XP_011533499.1 | |||
XM_011535198.1 | 7792 | UTR 3 | XP_011533500.1 | |||
XM_011535199.1 | 7792 | UTR 3 | XP_011533501.1 | |||
XM_011535200.2 | 7792 | UTR 3 | XP_011533502.1 | |||
XM_017020704.1 | 7792 | UTR 3 | XP_016876193.1 | |||
XM_017020705.1 | 7792 | UTR 3 | XP_016876194.1 | |||
XM_017020706.1 | 7792 | UTR 3 | XP_016876195.1 |