Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTAATGATTTCAGCTATACAT[T/A]ACATATACAACTCTGTAGCCTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENOX1 - ecto-NOX disulfide-thiol exchanger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127615.1 | 2913 | UTR 3 | NP_001121087.1 | |||
NM_001242863.1 | 2913 | UTR 3 | NP_001229792.1 | |||
NM_017993.3 | 2913 | UTR 3 | NP_060463.2 | |||
XM_005266439.3 | 2913 | UTR 3 | XP_005266496.1 | |||
XM_011535126.2 | 2913 | UTR 3 | XP_011533428.1 | |||
XM_011535127.2 | 2913 | UTR 3 | XP_011533429.1 | |||
XM_011535128.2 | 2913 | UTR 3 | XP_011533430.1 | |||
XM_011535132.2 | 2913 | UTR 3 | XP_011533434.1 | |||
XM_017020637.1 | 2913 | UTR 3 | XP_016876126.1 | |||
XM_017020638.1 | 2913 | UTR 3 | XP_016876127.1 | |||
XM_017020639.1 | 2913 | UTR 3 | XP_016876128.1 | |||
XM_017020640.1 | 2913 | UTR 3 | XP_016876129.1 | |||
XM_017020641.1 | 2913 | Intron | XP_016876130.1 | |||
XM_017020642.1 | 2913 | Intron | XP_016876131.1 |