Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCCCAGGCCTTGGTGCTACCTT[C/T]GCTGGCATCGTTCCAAGGGCTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LATS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LATS2 - large tumor suppressor kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014572.2 | 3526 | Missense Mutation | AAA,GAA | K,E 1016 | NP_055387.2 | |
XM_005266342.1 | 3526 | Missense Mutation | AAA,GAA | K,E 1016 | XP_005266399.1 | |
XM_011535042.2 | 3526 | Missense Mutation | AAA,GAA | K,E 929 | XP_011533344.1 | |
XM_017020541.1 | 3526 | Missense Mutation | AAA,GAA | K,E 909 | XP_016876030.1 | |
XM_017020542.1 | 3526 | UTR 3 | XP_016876031.1 |