Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGGTGACTGTGGAGGCAGATT[C/T]TCCCTAGCACATCTTCTCCGTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSC22D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSC22D1 - TSC22 domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243797.1 | 462 | UTR 3 | NP_001230726.1 | |||
NM_001243798.1 | 462 | UTR 3 | NP_001230727.1 | |||
NM_001243799.1 | 462 | Intron | NP_001230728.1 | |||
NM_006022.3 | 462 | UTR 3 | NP_006013.1 | |||
NM_183422.3 | 462 | UTR 3 | NP_904358.2 | |||
XM_005266583.4 | 462 | Intron | XP_005266640.1 | |||
XM_017020810.1 | 462 | Intron | XP_016876299.1 | |||
XM_017020811.1 | 462 | Intron | XP_016876300.1 |