Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTACCCTGAGAGTGGGTAGACT[G/A]AACAAAAACGGCGGGTGCACTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609239 MIM: 614041 | ||||||||||||||||||||
Literature Links: |
LPAR6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPAR6 - lysophosphatidic acid receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162497.1 | 2088 | Nonsense Mutation | CAG,TAG | Q,* 155 | NP_001155969.1 | |
NM_001162498.1 | 2088 | Nonsense Mutation | CAG,TAG | Q,* 155 | NP_001155970.1 | |
NM_005767.5 | 2088 | Nonsense Mutation | CAG,TAG | Q,* 155 | NP_005758.2 |
RB1 - RB transcriptional corepressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000321.2 | 2088 | Intron | NP_000312.2 | |||
XM_011535171.2 | 2088 | Intron | XP_011533473.1 |