Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATGGTTGTTCACACTCTGTTGA[A/G]GAGGGAGATAACTGAGAAAAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607531 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLF12 - Kruppel like factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007249.4 | 6607 | UTR 3 | NP_009180.3 | |||
XM_005266251.3 | 6607 | UTR 3 | XP_005266308.1 | |||
XM_011534907.2 | 6607 | UTR 3 | XP_011533209.1 | |||
XM_011534908.2 | 6607 | Intron | XP_011533210.1 | |||
XM_011534909.2 | 6607 | UTR 3 | XP_011533211.1 | |||
XM_011534910.2 | 6607 | UTR 3 | XP_011533212.1 | |||
XM_011534911.2 | 6607 | UTR 3 | XP_011533213.1 | |||
XM_011534912.2 | 6607 | UTR 3 | XP_011533214.1 | |||
XM_017020384.1 | 6607 | UTR 3 | XP_016875873.1 | |||
XM_017020385.1 | 6607 | UTR 3 | XP_016875874.1 |