Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACCAACGCGGAGGTCTCGGCGG[C/T]CGCTGCGGGAGCCACAGGAAGTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610041 | ||||||||||||||||||||
Literature Links: |
NDFIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDFIP2 - Nedd4 family interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161407.1 | 598 | Missense Mutation | GCC,GTC | A,V 39 | NP_001154879.1 | |
NM_019080.2 | 598 | Missense Mutation | GCC,GTC | A,V 39 | NP_061953.2 | |
XM_006719840.3 | 598 | Missense Mutation | GCC,GTC | A,V 39 | XP_006719903.1 |
NDFIP2-AS1 - NDFIP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |