Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATCTTCTTTTCTCTTTGTACTC[A/G]TAAACCAGAAGGCATCCTTAAGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR1D - RNA polymerase I subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206559.1 | Intron | NP_001193488.1 | ||||
NM_015972.3 | Intron | NP_057056.1 | ||||
NM_152705.2 | Intron | NP_689918.1 | ||||
XM_005266412.1 | Intron | XP_005266469.1 | ||||
XM_005266414.2 | Intron | XP_005266471.1 | ||||
XM_017020622.1 | Intron | XP_016876111.1 | ||||
XM_017020623.1 | Intron | XP_016876112.1 |