Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGATGAATAAAAAGGCACACT[A/C]GAAAAATTTGAGCGCAGAAAGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613218 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYB5A - cytochrome b5 type A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190807.2 | 702 | UTR 3 | NP_001177736.1 | |||
NM_001914.3 | 702 | UTR 3 | NP_001905.1 | |||
NM_148923.3 | 702 | UTR 3 | NP_683725.1 | |||
XM_011525835.2 | 702 | Intron | XP_011524137.1 |