Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGAACGCAGGCTGAGGTCGGAG[A/C]GCGCGGGCGTGGTGCGGGAGCTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604927 | ||||||||||||||||||||
Literature Links: |
CTDP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTDP1 - CTD phosphatase subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202504.1 | 406 | Intron | NP_001189433.1 | |||
NM_001318511.1 | 406 | Missense Mutation | AGC,CGC | S,R 87 | NP_001305440.1 | |
NM_004715.4 | 406 | Missense Mutation | AGC,CGC | S,R 87 | NP_004706.3 | |
NM_048368.3 | 406 | Missense Mutation | AGC,CGC | S,R 87 | NP_430255.2 | |
XM_011526261.1 | 406 | Missense Mutation | AGC,CGC | S,R 87 | XP_011524563.1 | |
XM_017026078.1 | 406 | Intron | XP_016881567.1 |
LOC105376875 - uncharacterized LOC105376875 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011526290.2 | 406 | Intron | XP_011524592.1 | |||
XM_011526292.2 | 406 | Intron | XP_011524594.1 | |||
XM_017026112.1 | 406 | Intron | XP_016881601.1 |
LOC284241 - uncharacterized LOC284241 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |