Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCTGCATCCGGATCTCCTGCC[C/T]TGGAGCGTACTCCTTGTCTCTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1468 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1468 - KIAA1468 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020854.3 | 113 | UTR 5 | NP_065905.2 | |||
XM_005266730.2 | 113 | UTR 5 | XP_005266787.1 | |||
XM_011526110.1 | 113 | UTR 5 | XP_011524412.1 | |||
XM_011526111.1 | 113 | UTR 5 | XP_011524413.1 | |||
XM_011526112.1 | 113 | UTR 5 | XP_011524414.1 | |||
XM_011526114.1 | 113 | UTR 5 | XP_011524416.1 | |||
XM_011526117.2 | 113 | Intron | XP_011524419.1 | |||
XM_017025882.1 | 113 | UTR 5 | XP_016881371.1 | |||
XM_017025883.1 | 113 | UTR 5 | XP_016881372.1 |
PIGN - phosphatidylinositol glycan anchor biosynthesis class N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012327.5 | 113 | Intron | NP_036459.1 | |||
NM_176787.4 | 113 | Intron | NP_789744.1 | |||
XM_011525889.1 | 113 | Intron | XP_011524191.1 | |||
XM_011525890.1 | 113 | Intron | XP_011524192.1 | |||
XM_011525891.1 | 113 | Intron | XP_011524193.1 | |||
XM_011525892.1 | 113 | Intron | XP_011524194.1 | |||
XM_011525893.1 | 113 | Intron | XP_011524195.1 | |||
XM_011525894.1 | 113 | Intron | XP_011524196.1 | |||
XM_011525895.1 | 113 | Intron | XP_011524197.1 | |||
XM_011525896.1 | 113 | Intron | XP_011524198.1 | |||
XM_011525898.1 | 113 | Intron | XP_011524200.1 | |||
XM_017025685.1 | 113 | Intron | XP_016881174.1 | |||
XM_017025686.1 | 113 | Intron | XP_016881175.1 |