Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCCCTCTTCTAGGCTAGCTC[G/T]CACTGGAGTCGCTGTGTTTCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602681 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXO3 - forkhead box O3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001455.3 | 390 | Intron | NP_001446.1 | |||
NM_201559.2 | 390 | Intron | NP_963853.1 | |||
XM_005266867.4 | 390 | Intron | XP_005266924.1 | |||
XM_005266868.3 | 390 | Intron | XP_005266925.1 | |||
XM_011535626.1 | 390 | Intron | XP_011533928.1 | |||
XM_011535628.2 | 390 | Intron | XP_011533930.1 | |||
XM_011535629.2 | 390 | Intron | XP_011533931.1 | |||
XM_017010585.1 | 390 | UTR 5 | XP_016866074.1 | |||
XM_017010586.1 | 390 | UTR 5 | XP_016866075.1 |