Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATGTAGAAAAAACAGCTGTTAT[C/G]AAGCACAAAATTTTAATCTAAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603356 | ||||||||||||||||||||
Literature Links: |
CD164 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD164 - CD164 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142401.1 | 1236 | UTR 3 | NP_001135873.1 | |||
NM_001142402.1 | 1236 | UTR 3 | NP_001135874.1 | |||
NM_001142403.1 | 1236 | Intron | NP_001135875.1 | |||
NM_001142404.1 | 1236 | Intron | NP_001135876.1 | |||
NM_006016.4 | 1236 | UTR 3 | NP_006007.2 |