Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCATACGAAGGACATGAATACC[A/G]AGAGACAAGAATCATTAAGAGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615358 MIM: 609390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK9 - adenylate kinase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FIG4 - FIG4 phosphoinositide 5-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014845.5 | Intron | NP_055660.1 | ||||
XM_011536281.2 | Intron | XP_011534583.1 | ||||
XM_017011591.1 | Intron | XP_016867080.1 | ||||
XM_017011592.1 | Intron | XP_016867081.1 | ||||
XM_017011593.1 | Intron | XP_016867082.1 |