Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGACAGTTGTACCGCAGCTTGT[C/T]TCTTTTCTAGGGCAGTGGGTCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180980 MIM: 614720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMD1 - adenosylmethionine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287214.1 | 655 | Intron | NP_001274143.1 | |||
NM_001287215.1 | 655 | Intron | NP_001274144.1 | |||
NM_001287216.1 | 655 | Intron | NP_001274145.1 | |||
NM_001634.5 | 655 | Intron | NP_001625.2 |
CDK19 - cyclin dependent kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300960.1 | 655 | Intron | NP_001287889.1 | |||
NM_001300963.1 | 655 | Intron | NP_001287892.1 | |||
NM_001300964.1 | 655 | UTR 5 | NP_001287893.1 | |||
NM_015076.4 | 655 | Intron | NP_055891.1 | |||
XM_005266871.3 | 655 | Intron | XP_005266928.1 | |||
XM_011535630.2 | 655 | Silent Mutation | GAA,GAG | E,E 96 | XP_011533932.1 | |
XM_011535631.2 | 655 | Silent Mutation | GAA,GAG | E,E 96 | XP_011533933.1 | |
XM_011535632.1 | 655 | Intron | XP_011533934.1 | |||
XM_017010587.1 | 655 | Intron | XP_016866076.1 | |||
XM_017010588.1 | 655 | Intron | XP_016866077.1 |