Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCTCGTCCAGCTGCGACTCAG[C/T]GCCTTCCACGTCCTGTTTGTATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607043 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF3IP2 - TRAF3 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164281.2 | 2647 | Missense Mutation | ACT,GCT | T,A 472 | NP_001157753.1 | |
NM_001164283.2 | 2647 | Missense Mutation | ACT,GCT | T,A 17 | NP_001157755.1 | |
NM_147200.2 | 2647 | Missense Mutation | ACT,GCT | T,A 482 | NP_671733.2 | |
NM_147686.3 | 2647 | Missense Mutation | ACT,GCT | T,A 473 | NP_679211.2 | |
XM_006715319.3 | 2647 | Missense Mutation | ACT,GCT | T,A 17 | XP_006715382.1 | |
XM_011535386.1 | 2647 | Missense Mutation | ACT,GCT | T,A 482 | XP_011533688.1 |
TRAF3IP2-AS1 - TRAF3IP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |