Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATTTGGGTAAGAAATGAGTGAA[C/T]AGGAATGCTTATAAGAGGTAGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM229B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM229B - family with sequence similarity 229 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LAMA4 - laminin subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105206.2 | Intron | NP_001098676.2 | ||||
NM_001105207.2 | Intron | NP_001098677.2 | ||||
NM_001105208.2 | Intron | NP_001098678.1 | ||||
NM_001105209.2 | Intron | NP_001098679.1 | ||||
NM_002290.4 | Intron | NP_002281.3 | ||||
XM_005266983.3 | Intron | XP_005267040.2 | ||||
XM_005266984.3 | Intron | XP_005267041.2 | ||||
XM_017010854.1 | Intron | XP_016866343.1 |