Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCCAAGACACTAGGCAGGCAG[C/T]GAAACCACAAACACACAGCAAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN1A1 - mannosidase alpha class 1A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005907.3 | Intron | NP_005898.2 | ||||
XM_005266986.4 | Intron | XP_005267043.1 | ||||
XM_011535833.2 | Intron | XP_011534135.1 | ||||
XM_011535834.2 | Intron | XP_011534136.1 |