Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGAGGAATTTGAAGCCAATAC[C/T]ATGGATTCTCTGGTAGACATGCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604693 | ||||||||||||||||||||
Literature Links: |
AKAP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP7 - A-kinase anchoring protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004842.3 | 126 | Intron | NP_004833.1 | |||
NM_016377.3 | 126 | Silent Mutation | ACC,ACT | T,T 31 | NP_057461.2 | |
NM_138633.2 | 126 | Intron | NP_619539.1 | |||
XM_005267228.1 | 126 | Silent Mutation | ACC,ACT | T,T 31 | XP_005267285.1 | |
XM_005267229.4 | 126 | Silent Mutation | ACC,ACT | T,T 9 | XP_005267286.1 | |
XM_006715614.2 | 126 | Silent Mutation | ACC,ACT | T,T 31 | XP_006715677.1 | |
XM_011536262.1 | 126 | Silent Mutation | ACC,ACT | T,T 31 | XP_011534564.1 | |
XM_017011506.1 | 126 | Silent Mutation | ACC,ACT | T,T 31 | XP_016866995.1 | |
XM_017011507.1 | 126 | Silent Mutation | ACC,ACT | T,T 31 | XP_016866996.1 | |
XM_017011508.1 | 126 | Silent Mutation | ACC,ACT | T,T 41 | XP_016866997.1 | |
XM_017011509.1 | 126 | Silent Mutation | ACC,ACT | T,T 35 | XP_016866998.1 | |
XM_017011510.1 | 126 | Intron | XP_016866999.1 | |||
XM_017011511.1 | 126 | Intron | XP_016867000.1 |