Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCAGGGAAGATAACCCTTCATA[A/C]TGAGAACCTTGCTCAGGTTGCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EYA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EYA4 - EYA transcriptional coactivator and phosphatase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301012.1 | Intron | NP_001287941.1 | ||||
NM_001301013.1 | Intron | NP_001287942.1 | ||||
NM_004100.4 | Intron | NP_004091.3 | ||||
NM_172103.3 | Intron | NP_742101.2 | ||||
NM_172105.3 | Intron | NP_742103.1 | ||||
XM_005266851.4 | Intron | XP_005266908.1 | ||||
XM_005266853.4 | Intron | XP_005266910.1 | ||||
XM_017010368.1 | Intron | XP_016865857.1 | ||||
XM_017010369.1 | Intron | XP_016865858.1 | ||||
XM_017010370.1 | Intron | XP_016865859.1 | ||||
XM_017010371.1 | Intron | XP_016865860.1 | ||||
XM_017010372.1 | Intron | XP_016865861.1 | ||||
XM_017010373.1 | Intron | XP_016865862.1 | ||||
XM_017010374.1 | Intron | XP_016865863.1 | ||||
XM_017010375.1 | Intron | XP_016865864.1 |