Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTCTTATTTCTGCAGCATATAT[A/G]GCAGTGACGAGGATGATGAGGACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189990 | ||||||||||||||||||||
Literature Links: |
MYB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYB - MYB proto-oncogene, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130172.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001123644.1 | |
NM_001130173.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001123645.1 | |
NM_001161656.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001155128.1 | |
NM_001161657.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001155129.1 | |
NM_001161658.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001155130.1 | |
NM_001161659.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001155131.1 | |
NM_001161660.1 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_001155132.1 | |
NM_005375.3 | 230 | Missense Mutation | AGC,GGC | S,G 11 | NP_005366.2 |