Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTATAGAGCATTCCTACCACTGT[C/T]TGAACTGACAGACCATCTGAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605196 | ||||||||||||||||||||
Literature Links: |
COQ3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COQ3 - coenzyme Q3, methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017421.3 | 933 | Silent Mutation | CAA,CAG | Q,Q 302 | NP_059117.3 | |
XM_005267018.4 | 933 | Silent Mutation | CAA,CAG | Q,Q 221 | XP_005267075.1 | |
XM_006715500.3 | 933 | Silent Mutation | CAA,CAG | Q,Q 343 | XP_006715563.1 | |
XM_011535898.2 | 933 | Silent Mutation | CAA,CAG | Q,Q 262 | XP_011534200.1 | |
XM_011535899.1 | 933 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_011534201.1 | |
XM_017010933.1 | 933 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_016866422.1 |