Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGTTCTTGTGATTATCTTCAT[A/G]TCTCTAAATCATATGCTTAAACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCMT1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252049.1 | Intron | NP_001238978.1 | ||||
NM_001252050.1 | Intron | NP_001238979.1 | ||||
NM_001252051.1 | Intron | NP_001238980.1 | ||||
NM_001252052.1 | Intron | NP_001238981.1 | ||||
NM_001252053.1 | Intron | NP_001238982.1 | ||||
NM_005389.2 | Intron | NP_005380.2 | ||||
XM_011535868.2 | Intron | XP_011534170.1 |