Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTCCCGGGAGGACTCGGCGCC[C/T]GTGGCCACGGCGGCCGCTGCAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613242 | ||||||||||||||||||||
Literature Links: |
PPP1R14C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R14C - protein phosphatase 1 regulatory inhibitor subunit 14C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030949.2 | 288 | Silent Mutation | CCC,CCT | P,P 49 | NP_112211.1 | |
XM_005267152.4 | 288 | Intron | XP_005267209.1 |