Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCTATTTTATTTAACTGTTTT[A/G]CTTTGTTAGCTTGATTTAAAACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNJ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYNJ2 - synaptojanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178088.1 | Intron | NP_001171559.1 | ||||
NM_003898.3 | Intron | NP_003889.1 | ||||
XM_005267198.4 | Intron | XP_005267255.1 | ||||
XM_005267199.2 | Intron | XP_005267256.1 | ||||
XM_005267200.1 | Intron | XP_005267257.1 | ||||
XM_006715592.3 | Intron | XP_006715655.1 | ||||
XM_011536224.1 | Intron | XP_011534526.1 | ||||
XM_011536225.1 | Intron | XP_011534527.1 | ||||
XM_011536226.2 | Intron | XP_011534528.1 | ||||
XM_011536227.2 | Intron | XP_011534529.1 | ||||
XM_011536228.2 | Intron | XP_011534530.1 | ||||
XM_011536230.2 | Intron | XP_011534532.1 | ||||
XM_017011406.1 | Intron | XP_016866895.1 |