Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCTGAGTTCTTTGACTTCCAA[C/T]GAGGGGAAGAGAAGATAGCGAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERAC1 - serine active site containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032861.3 | 1726 | Silent Mutation | TCA,TCG | S,S 553 | NP_116250.3 | |
XM_006715586.2 | 1726 | Silent Mutation | TCA,TCG | S,S 483 | XP_006715649.1 | |
XM_011536196.2 | 1726 | Silent Mutation | TCA,TCG | S,S 546 | XP_011534498.1 | |
XM_011536198.2 | 1726 | Silent Mutation | TCA,TCG | S,S 483 | XP_011534500.1 |