Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTAATTTGGGGCATTAGCTGCC[A/G]CAATATCTTATATCTCTTAAGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUT9 - fucosyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006581.3 | Intron | NP_006572.2 | ||||
XM_011535383.2 | Intron | XP_011533685.1 | ||||
XM_011535385.2 | Intron | XP_011533687.1 | ||||
XM_017010188.1 | Intron | XP_016865677.1 | ||||
XM_017010190.1 | Intron | XP_016865679.1 |