Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTCTTTTCCACATTACGTCCTT[A/G]GACTTCAGGCATAACTCCAAGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602544 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARK2 - parkin RBR E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004562.2 | Intron | NP_004553.2 | ||||
NM_013987.2 | Intron | NP_054642.2 | ||||
NM_013988.2 | Intron | NP_054643.2 | ||||
XM_011535863.1 | Intron | XP_011534165.1 | ||||
XM_017010908.1 | Intron | XP_016866397.1 | ||||
XM_017010909.1 | Intron | XP_016866398.1 |