Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGAGCCCTCGACCCTGCTCACC[A/G]GCTCCTCTGGCTGCTGCCGTCTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187020 | |||||||||||||||||||||||||||||||||||
Literature Links: |
TCP10 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
AMR
|
|||||
EUR - Not Available |
TCP10 - t-complex 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004610.3 | 753 | Intron | NP_004601.3 | |||
XM_011536082.2 | 753 | Intron | XP_011534384.1 | |||
XM_011536084.2 | 753 | Intron | XP_011534386.1 | |||
XM_011536085.2 | 753 | Intron | XP_011534387.1 | |||
XM_011536088.2 | 753 | Intron | XP_011534390.1 | |||
XM_011536090.2 | 753 | Intron | XP_011534392.1 | |||
XM_017011228.1 | 753 | Intron | XP_016866717.1 | |||
XM_017011229.1 | 753 | Intron | XP_016866718.1 | |||
XM_017011230.1 | 753 | Intron | XP_016866719.1 | |||
XM_017011231.1 | 753 | Silent Mutation | GCC,GCT | A,A 229 | XP_016866720.1 |