Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTCTGCCTATAAGGAATAATAC[A/C]TAATTTTAGATATAAAGGCCCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600866 MIM: 600075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDCD2 - programmed cell death 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199461.1 | 2744 | Intron | NP_001186390.1 | |||
NM_001199462.1 | 2744 | UTR 3 | NP_001186391.1 | |||
NM_001199463.1 | 2744 | Intron | NP_001186392.1 | |||
NM_001199464.1 | 2744 | Intron | NP_001186393.1 | |||
NM_002598.3 | 2744 | UTR 3 | NP_002589.2 | |||
NM_144781.2 | 2744 | Intron | NP_659005.1 | |||
XM_017010926.1 | 2744 | Intron | XP_016866415.1 |
TBP - TATA-box binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |