Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGCTTGGCTTGGAGCCACCACA[C/T]GCAACAGCATGAAACTTTCTCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BVES PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BVES - blood vessel epicardial substance | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199563.1 | 4752 | UTR 3 | NP_001186492.1 | |||
NM_007073.4 | 4752 | UTR 3 | NP_009004.2 | |||
NM_147147.3 | 4752 | UTR 3 | NP_671488.1 | |||
XM_011535398.2 | 4752 | Intron | XP_011533700.1 |