Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCTAGCTTCAGCACACTTGCTA[G/T]TAGCAGCTACTTCTTTTTCACATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DTD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTD2 - D-tyrosyl-tRNA deacylase 2 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080664.2 | 479 | Missense Mutation | AAT,ACT | N,T 133 | NP_542395.1 |
LOC101927124 - uncharacterized LOC101927124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |