Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGAAGCTTTGGAAGAGCCCATAC[A/C]TTGGGCTAGGCCCAGGGCACTCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609947 MIM: 615902 | ||||||||||||||||||||
Literature Links: |
KIAA0391 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0391 - KIAA0391 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256678.1 | 439 | Missense Mutation | ATT,CTT | I,L 20 | NP_001243607.1 | |
NM_001256679.1 | 439 | Intron | NP_001243608.1 | |||
NM_001256680.1 | 439 | Intron | NP_001243609.1 | |||
NM_001256681.1 | 439 | Intron | NP_001243610.1 | |||
NM_014672.3 | 439 | Missense Mutation | ATT,CTT | I,L 20 | NP_055487.2 | |
XM_005268237.3 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_005268294.1 | |
XM_011537409.2 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_011535711.1 | |
XM_011537410.2 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_011535712.1 | |
XM_017021835.1 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_016877324.1 | |
XM_017021836.1 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_016877325.1 | |
XM_017021837.1 | 439 | Missense Mutation | ATT,CTT | I,L 20 | XP_016877326.1 |
PPP2R3C - protein phosphatase 2 regulatory subunit B''gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305155.1 | 439 | Intron | NP_001292084.1 | |||
NM_001305156.1 | 439 | Intron | NP_001292085.1 | |||
NM_017917.3 | 439 | Intron | NP_060387.2 | |||
XM_005267782.3 | 439 | Intron | XP_005267839.1 | |||
XM_017021388.1 | 439 | Intron | XP_016876877.1 | |||
XM_017021389.1 | 439 | Intron | XP_016876878.1 |