Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTATATGAGGCCAAGGGGTGA[A/G]AACACACATACACACACCCTGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609431 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MBIP - MAP3K12 binding inhibitory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144891.1 | Intron | NP_001138363.1 | ||||
NM_001308110.1 | Intron | NP_001295039.1 | ||||
NM_016586.2 | Intron | NP_057670.2 | ||||
XM_005267754.3 | Intron | XP_005267811.1 | ||||
XM_005267756.4 | Intron | XP_005267813.1 | ||||
XM_017021367.1 | Intron | XP_016876856.1 |