Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACGTTCCAGATGTTTTGGGCAT[G/A]TTCTTTAATTGTTCGGTCACTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613741 | ||||||||||||||||||||
Literature Links: |
ABHD12B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD12B - abhydrolase domain containing 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206673.1 | 2486 | Intron | NP_001193602.1 | |||
NM_181533.3 | 2486 | Intron | NP_853511.2 | |||
NM_181814.1 | 2486 | Intron | NP_861535.1 | |||
XM_006720052.1 | 2486 | Intron | XP_006720115.1 | |||
XM_011536472.1 | 2486 | Intron | XP_011534774.1 | |||
XM_011536474.2 | 2486 | Intron | XP_011534776.1 | |||
XM_017021031.1 | 2486 | Intron | XP_016876520.1 |
PYGL - phosphorylase, glycogen, liver | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163940.1 | 2486 | Missense Mutation | CAT,TAT | H,Y 787 | NP_001157412.1 | |
NM_002863.4 | 2486 | Missense Mutation | CAT,TAT | H,Y 821 | NP_002854.3 |