Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATCCCCAGATTTCTTCATATGC[C/T]CAAAAGATTCTGTGGAAATTGGATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616510 MIM: 615814 | ||||||||||||||||||||
Literature Links: |
GNPNAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GNPNAT1 - glucosamine-phosphate N-acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198066.3 | 516 | Missense Mutation | GAG,GGG | E,G 76 | NP_932332.1 | |
XM_005268012.2 | 516 | Missense Mutation | GAG,GGG | E,G 76 | XP_005268069.1 | |
XM_006720238.3 | 516 | Missense Mutation | GAG,GGG | E,G 76 | XP_006720301.1 |
STYX - serine/threonine/tyrosine interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |