Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAATCAAGTCAGTCATTTCCT[A/G]TGCCTGTTTCTAAGCTTTGAAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35F4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35F4 - solute carrier family 35 member F4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206920.1 | Intron | NP_001193849.1 | ||||
NM_001306087.1 | Intron | NP_001293016.1 | ||||
XM_011536720.2 | Intron | XP_011535022.1 | ||||
XM_011536721.2 | Intron | XP_011535023.1 | ||||
XM_011536723.2 | Intron | XP_011535025.1 | ||||
XM_011536724.2 | Intron | XP_011535026.1 | ||||
XM_011536725.1 | Intron | XP_011535027.1 | ||||
XM_017021258.1 | Intron | XP_016876747.1 | ||||
XM_017021259.1 | Intron | XP_016876748.1 | ||||
XM_017021260.1 | Intron | XP_016876749.1 | ||||
XM_017021261.1 | Intron | XP_016876750.1 |