Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTCCTTTGCTTTACTCTGCAA[A/G]GAAGTATAGTAGAGCTGTGATTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MNAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MNAT1 - MNAT1, CDK activating kinase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177963.1 | Intron | NP_001171434.1 | ||||
NM_002431.3 | Intron | NP_002422.1 | ||||
XM_005267687.2 | Intron | XP_005267744.1 | ||||
XM_005267688.2 | Intron | XP_005267745.1 | ||||
XM_017021332.1 | Intron | XP_016876821.1 | ||||
XM_017021333.1 | Intron | XP_016876822.1 | ||||
XM_017021334.1 | Intron | XP_016876823.1 |