Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGACAGTACCAACTAAATTTC[C/T]TCATTCCCTTGTCCTATTCCCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601647 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R5E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP2R5E - protein phosphatase 2 regulatory subunit B'epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282179.1 | Intron | NP_001269108.1 | ||||
NM_001282180.1 | Intron | NP_001269109.1 | ||||
NM_001282181.1 | Intron | NP_001269110.1 | ||||
NM_001282182.1 | Intron | NP_001269111.1 | ||||
NM_006246.3 | Intron | NP_006237.1 |