Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGAATATGTATTTCAGTATTTC[C/T]GAGTAACCCGTCACTTCAAAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPA2 - heat shock protein family A (Hsp70) member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R36 - protein phosphatase 1 regulatory subunit 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172365.2 | Intron | NP_758953.1 | ||||
XM_005267354.3 | Intron | XP_005267411.1 | ||||
XM_005267355.3 | Intron | XP_005267412.1 | ||||
XM_011536468.2 | Intron | XP_011534770.1 |