Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTAGACTTCACCAGGGAAGCC[G/T]TCTAGTCCTGGGCTTTTCTTCATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PLEKHG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLEKHG3 - pleckstrin homology and RhoGEF domain containing G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308147.1 | Intron | NP_001295076.1 | ||||
XM_011536624.2 | Intron | XP_011534926.1 | ||||
XM_011536625.2 | Intron | XP_011534927.1 | ||||
XM_011536627.2 | Intron | XP_011534929.1 | ||||
XM_011536628.2 | Intron | XP_011534930.1 | ||||
XM_011536629.2 | Intron | XP_011534931.1 | ||||
XM_017021154.1 | Intron | XP_016876643.1 | ||||
XM_017021155.1 | Intron | XP_016876644.1 | ||||
XM_017021156.1 | Intron | XP_016876645.1 | ||||
XM_017021157.1 | Intron | XP_016876646.1 | ||||
XM_017021158.1 | Intron | XP_016876647.1 |