Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGGCGGCGGCAGCAGCAGCGG[C/G]GGCTGCGGGCGGTCGGGGCTCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601931 MIM: 602279 | ||||||||||||||||||||
Literature Links: |
BCL2L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL2L2 - BCL2 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BCL2L2-PABPN1 - BCL2L2-PABPN1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199864.1 | 1317 | Intron | NP_001186793.1 |
PABPN1 - poly(A) binding protein nuclear 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004643.3 | 1317 | Missense Mutation | GCG,GGG | A,G 12 | NP_004634.1 |